The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177878.1 | NP_001171349.1 | 8 - 9 | 1256 | 78 |
NM_001177879.1 | NP_001171350.1 | 9 - 10 | 1070 | 78 | |
NM_172783.3 | NP_766371.1 | 8 - 9 | 1256 | 78 | |
GenBank mRNA | AK052346.1 | - | 8 - 9 | 1001 | 78 |
AK052992.1 | - | 8 - 9 | 1257 | 78 | |
AK165506.1 | - | 9 - 10 | 1071 | 78 | |
BC050040.1 | - | 9 - 10 | 1029 | 78 | |
BC051167.1 | - | 3 - 4 | 496 | 78 | |
BC058254.1 | - | 8 - 9 | 1225 | 78 |