The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172785.3 | NP_766373.2 | 1 - 2 | 91 | 73 |
GenBank mRNA | AK052806.1 | - | 1 - 2 | 92 | 73 |
BB661745.1 | - | 1 - 2 | 92 | 73 |