The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253759.1 | NP_001240688.1 | 15 - 16 | 2253 | 93 |
NM_172813.3 | NP_766401.1 | 14 - 15 | 2196 | 93 | |
GenBank mRNA | AK045500.1 | - | 13 - 14 | 2007 | 93 |
AK051821.1 | - | 14 - 15 | 2144 | 93 | |
AK082388.1 | - | 13 - 14 | 2090 | 93 | |
BC052062.1 | - | 12 - 13 | 1764 | 93 |