The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172857.2 | NP_766445.1 | 3 - 4 | 902 | 63 |
GenBank mRNA | AK030132.1 | - | 3 - 4 | 904 | 63 |
BC092241.1 | - | 1 - 2 | 156 | 63 | |
BC144750.1 | - | 2 - 3 | 285 | 63 | |
BC150662.1 | - | 3 - 4 | 379 | 63 |