The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172866.3 | NP_766454.1 | 6 - 7 | 923 | 58 |
GenBank mRNA | AK089288.1 | - | 6 - 7 | 706 | 58 |
AK129103.1 | - | 6 - 7 | 709 | 58 | |
BC086614.1 | - | 6 - 7 | 729 | 58 |