The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172913.3 | NP_766501.2 | 2 - 3 | 453 | 92 |
GenBank mRNA | AK082602.1 | - | 2 - 3 | 451 | 92 |
BC052044.1 | - | 2 - 3 | 179 | 92 | |
CK619337.1 | - | 2 - 3 | 544 | 92 | |
EF410108.1 | - | - | 317 | 92 |