Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf3
Entrez Gene ID
245525
Gene Name
heat shock transcription factor 3
Gene Aliases
B230358A15Rik, HSF 3, HSTF 3, mHSF3
Chromosome Location
Chr.X: 96306184 - 96367919 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172931.3 NP_766519.1 3 - 4 358 104
GenBank mRNA AK046229.1 - - 388 104
BC117805.2 - 3 - 4 358 104

View Full Product Details