The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172960.3 | NP_766548.2 | 13 - 14 | 1649 | 57 |
NR_073123.1 | 13 - 14 | 1644 | 57 | ||
GenBank mRNA | AK039217.1 | - | 13 - 14 | 1645 | 57 |
AK163557.1 | - | 12 - 13 | 1676 | 57 | |
BC030881.1 | - | 10 - 11 | 1872 | 57 | |
BC053696.1 | - | 3 - 4 | 837 | 57 | |
BC060528.1 | - | 13 - 14 | 1533 | 57 |