The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173185.2 | NP_775277.2 | 13 - 14 | 1746 | 96 |
GenBank mRNA | AK090262.1 | - | 13 - 14 | 1747 | 96 |