The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173385.2 | NP_775561.1 | 5 - 6 | 597 | 94 |
GenBank mRNA | AK036473.1 | - | 5 - 6 | 597 | 94 |
AK081544.1 | - | 5 - 6 | 598 | 94 | |
BC058167.1 | - | 5 - 6 | 680 | 94 | |
BC080666.1 | - | 4 - 5 | 593 | 94 |