The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175092.3 | NP_780301.1 | 4 - 5 | 723 | 85 |
GenBank mRNA | AK038837.1 | - | - | 491 | 85 |
AK172212.1 | - | 4 - 5 | 491 | 85 | |
BC096597.1 | - | 4 - 5 | 513 | 85 | |
BC132355.1 | - | - | 555 | 85 | |
BC132357.1 | - | - | 555 | 85 | |
DV646753.1 | - | 1 - 2 | 145 | 85 |