The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011120.2 | NP_035250.1 | 4 - 5 | 419 | 92 |
GenBank mRNA | AK005421.1 | - | 4 - 5 | 416 | 92 |
AK005557.1 | - | 4 - 5 | 419 | 92 | |
AK019461.1 | - | 4 - 5 | 419 | 92 | |
BC099470.1 | - | 4 - 5 | 408 | 92 | |
X02594.1 | - | 4 - 5 | 419 | 92 |