Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim10
Entrez Gene ID
19824
Gene Name
tripartite motif-containing 10
Gene Aliases
AI324236, BB139825, Herf1, Rnf9
Chromosome Location
Chr.17: 36869574 - 36877833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011280.2 NP_035410.2 2 - 3 829 110
GenBank mRNA AF134811.1 - 2 - 3 811 110
AF220121.1 - 2 - 3 816 110
AK011082.1 - 2 - 3 829 110
BC051632.1 - 2 - 3 802 110

View Full Product Details