The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011280.2 | NP_035410.2 | 3 - 4 | 1057 | 89 |
GenBank mRNA | AF134811.1 | - | 3 - 4 | 1039 | 89 |
AF220121.1 | - | 3 - 4 | 1044 | 89 | |
AK011082.1 | - | 3 - 4 | 1057 | 89 | |
BC051632.1 | - | 3 - 4 | 1030 | 89 |