Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tpsg1
Entrez Gene ID
26945
Gene Name
tryptase gamma 1
Gene Aliases
Prss31, Tmt
Chromosome Location
Chr.17: 25369216 - 25374442 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012034.3 NP_036164.2 3 - 4 585 79
GenBank mRNA AF175523.1 - 3 - 4 486 79
AK027982.1 - 2 - 3 411 79
BC019974.1 - 3 - 4 509 79
BC052325.1 - 3 - 4 554 79

View Full Product Details