Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyar
Entrez Gene ID
17089
Gene Name
Ly1 antibody reactive clone
Gene Aliases
MLZ-264
Chromosome Location
Chr.5: 38220482 - 38234306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025281.3 NP_079557.2 3 - 4 438 76
GenBank mRNA AK006373.1 - 3 - 4 372 76
AK076727.1 - 3 - 4 443 76
BC116923.1 - 2 - 3 294 76
BC116927.1 - 2 - 3 294 76
S60885.1 - 3 - 4 385 76

View Full Product Details