The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080726.3 | NP_542764.2 | 3 - 4 | 697 | 81 |
GenBank mRNA | AK077317.1 | - | 3 - 4 | 578 | 81 |
AK090165.1 | - | 3 - 4 | 698 | 81 | |
BC018219.1 | - | 3 - 4 | 579 | 81 |