The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175236.4 | NP_780445.1 | 10 - 11 | 1029 | 107 |
NR_027664.1 | 9 - 10 | 988 | 107 | ||
GenBank mRNA | AK018241.1 | - | 10 - 11 | 1000 | 107 |
AK038853.1 | - | 10 - 11 | 996 | 107 | |
AK046780.1 | - | 10 - 11 | 955 | 107 | |
AK050178.1 | - | 9 - 10 | 953 | 107 | |
AK084104.1 | - | - | 958 | 107 | |
BC026584.1 | - | 10 - 11 | 987 | 107 |