Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttyh3
Entrez Gene ID
78339
Gene Name
tweety homolog 3 (Drosophila)
Gene Aliases
2900029G13Rik, AI414930, TTY3, mTTY3
Chromosome Location
Chr.5: 140620578 - 140649031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK052340.1 - 13 - 14 1695 105
AK173233.1 - 10 - 11 1084 105
BC062917.1 - 13 - 14 1701 105

View Full Product Details