The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175324.3 | NP_780533.2 | 11 - 12 | 1468 | 104 |
GenBank mRNA | AK077531.1 | - | 11 - 12 | 1469 | 104 |
AK128998.1 | - | 11 - 12 | 1447 | 104 | |
AK139918.1 | - | 11 - 12 | 1412 | 104 | |
AK145561.1 | - | 11 - 12 | 1457 | 104 | |
AK145834.1 | - | 11 - 12 | 1448 | 104 | |
BC006042.1 | - | 1 - 2 | 144 | 104 | |
BC032994.1 | - | 4 - 5 | 397 | 104 | |
BC045199.1 | - | 11 - 12 | 1432 | 104 |