The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177368.4 | NP_796342.2 | 2 - 3 | 1382 | 128 |
GenBank mRNA | AK052344.1 | - | 2 - 3 | 1355 | 128 |
BC090655.1 | - | 2 - 3 | 743 | 129 | |
BC092226.1 | - | 2 - 3 | 1338 | 128 | |
BC138362.1 | - | 2 - 3 | 1113 | 128 | |
BC138363.1 | - | 2 - 3 | 1113 | 128 |