The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177546.2 | NP_808214.1 | 5 - 6 | 798 | 73 |
NM_211138.1 | NP_997593.1 | 5 - 6 | 809 | 73 | |
GenBank mRNA | AK032027.1 | - | 5 - 6 | 511 | 73 |
AK053577.1 | - | 5 - 6 | 783 | 73 | |
AK149304.1 | - | 5 - 6 | 799 | 73 | |
AY189949.1 | - | 5 - 6 | 646 | 73 | |
AY189950.1 | - | 5 - 6 | 798 | 73 | |
BC048917.1 | - | 5 - 6 | 680 | 73 |