The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177580.3 | NP_808248.1 | 4 - 5 | 398 | 59 |
GenBank mRNA | AK143783.1 | - | 4 - 5 | 383 | 59 |
BC048937.1 | - | 4 - 5 | 336 | 59 |