Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekhh2
Entrez Gene ID
213556
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 2
Gene Aliases
AI256725, E030001K05, mKIAA2028
Chromosome Location
Chr.17: 84511895 - 84622142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177606.4 NP_808274.2 8 - 9 1731 86
GenBank mRNA AK029252.1 - 8 - 9 1733 86
AK173337.1 - 5 - 6 1387 86
BC021518.1 - 8 - 9 1703 86
BC125583.1 - 8 - 9 1696 86
BC137804.1 - 8 - 9 1693 86

View Full Product Details