Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
214580
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
5430423O14Rik, 5730458D16Rik, Gm1099, R75284
Chromosome Location
Chr.7: 131371113 - 131392768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039534.1 NP_001034623.1 3 - 4 739 82
GenBank mRNA AI173292.1 - 1 - 2 235 82
AK077585.1 - 3 - 4 740 82
BC030313.1 - 1 - 2 1231 82
BC110381.1 - 3 - 4 715 82
BC115503.2 - 3 - 4 740 82

View Full Product Details