The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177615.3 | NP_808283.2 | 17 - 18 | 2063 | 63 |
GenBank mRNA | AK052641.1 | - | - | 2417 | 63 |
BC089587.1 | - | 16 - 17 | 2415 | 63 |