The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178372.2 | NP_848459.1 | 3 - 4 | 634 | 131 |
GenBank mRNA | AK081986.1 | - | 3 - 4 | 636 | 131 |
AY261775.1 | - | 3 - 4 | 634 | 131 | |
BC109366.2 | - | 3 - 4 | 608 | 131 |