The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178372.2 | NP_848459.1 | 4 - 5 | 791 | 76 |
GenBank mRNA | AK081986.1 | - | 4 - 5 | 793 | 76 |
AY261775.1 | - | 4 - 5 | 791 | 76 | |
BC109366.2 | - | 4 - 5 | 765 | 76 |