Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab3gap1
Entrez Gene ID
226407
Gene Name
RAB3 GTPase activating protein subunit 1
Gene Aliases
1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130
Chromosome Location
Chr.1: 127868737 - 127943876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178690.4 NP_848805.2 1 - 2 59 70
GenBank mRNA AK035603.1 - 1 - 2 43 70
AK076399.1 - 1 - 2 42 70
AK080432.1 - 1 - 2 41 70
AK134604.1 - 1 - 2 41 70
AK164523.1 - 1 - 2 40 70
AK172881.1 - 1 - 2 46 70
CJ084350.1 - 1 - 2 60 70

View Full Product Details