The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038624.1 | NP_001033713.1 | 3 - 4 | 463 | 124 |
NM_178780.3 | NP_848895.2 | 3 - 4 | 463 | 124 | |
GenBank mRNA | AK053760.1 | - | 3 - 4 | 444 | 124 |
AK082275.1 | - | 3 - 4 | 444 | 124 | |
AK134663.1 | - | 3 - 4 | 445 | 124 | |
AK138641.1 | - | 3 - 4 | 462 | 124 | |
AK149334.1 | - | 3 - 4 | 451 | 124 | |
BC059258.1 | - | 2 - 3 | 205 | 124 |