Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcun1d4
Entrez Gene ID
100737
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae)
Gene Aliases
AI836376
Chromosome Location
Chr.5: 73481027 - 73560794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190733.1 NP_001177662.1 5 - 6 481 65
NM_001190734.1 NP_001177663.1 6 - 7 416 65
NM_178896.5 NP_849227.1 5 - 6 481 65
GenBank mRNA AK033559.1 - 5 - 6 426 65
AK076991.1 - 6 - 7 417 65
BC060213.1 - 5 - 6 423 65
BC108346.1 - 5 - 6 383 65
BC137627.1 - 5 - 6 425 65
BC137628.1 - 5 - 6 425 65
BC144794.1 - 5 - 6 425 65
CO433728.1 - 1 - 2 90 65

View Full Product Details