Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam69c
Entrez Gene ID
240479
Gene Name
family with sequence similarity 69, member C
Gene Aliases
B230399E16Rik
Chromosome Location
Chr.18: 84718987 - 84741731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173770.4 NP_776131.1 2 - 3 262 71
GenBank mRNA AK046504.1 - 2 - 3 263 71

View Full Product Details