Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Celf5
Entrez Gene ID
319586
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
4930565A21Rik, Brunol5
Chromosome Location
Chr.10: 81459227 - 81482725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176954.3 NP_795928.2 4 - 5 510 69
GenBank mRNA AK029796.1 - 4 - 5 1684 69
AK083461.1 - 3 - 4 238 69
AK132321.1 - 4 - 5 476 69
AK158669.1 - 3 - 4 287 69
BC079845.1 - 4 - 5 482 69

View Full Product Details