Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Celf5
Entrez Gene ID
319586
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
4930565A21Rik, Brunol5
Chromosome Location
Chr.10: 81459227 - 81482725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176954.3 NP_795928.2 5 - 6 593 77
GenBank mRNA AK029796.1 - 5 - 6 1767 77
AK083461.1 - 4 - 5 321 77
AK132321.1 - 5 - 6 559 77
AK158669.1 - 4 - 5 370 77
BC079845.1 - 5 - 6 565 77

View Full Product Details