Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl7
Entrez Gene ID
448987
Gene Name
F-box and leucine-rich repeat protein 7
Gene Aliases
AL023057, D230018M15Rik, FBL7, Fbl6
Chromosome Location
Chr.15: 26540454 - 26895564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK084174.1 - 1 - 1 1004 80
AK084297.1 - 1 - 1 458 80
BC050864.1 - 2 - 2 1511 80

View Full Product Details