Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nt5dc1
Entrez Gene ID
319638
Gene Name
5'-nucleotidase domain containing 1
Gene Aliases
6030401B09Rik, AW987726, Nt5c2l1
Chromosome Location
Chr.10: 34303609 - 34418589 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176968.4 NP_795942.2 4 - 5 468 70
GenBank mRNA AK049656.1 - 4 - 5 468 70
AK077876.1 - 4 - 5 469 70
AK145666.1 - 4 - 5 469 70
AK161350.1 - 4 - 5 422 70
AK164144.1 - 4 - 5 412 70
AK171761.1 - 4 - 5 481 70
BC098476.1 - 4 - 5 431 70
BC125276.1 - 4 - 5 467 70

View Full Product Details