Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vwc2
- Entrez Gene ID
-
319922
- Gene Name
- von Willebrand factor C domain containing 2
- Gene Aliases
-
A930041G11Rik, G11, PSST739, UNQ739, cradin
- Chromosome Location
-
Chr.11: 11113859 - 11269295 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK044771.1 |
- |
1 - 2 |
34 |
79 |
View Full Product Details