The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177091.5 | NP_796065.2 | 5 - 6 | 1162 | 72 |
GenBank mRNA | AK053989.1 | - | 5 - 6 | 1025 | 72 |
AK054241.1 | - | 5 - 6 | 1162 | 72 | |
AK165075.1 | - | 2 - 3 | 717 | 72 | |
BC113188.1 | - | 5 - 6 | 968 | 72 |