The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177723.4 | NP_808391.2 | 1 - 2 | 179 | 64 |
GenBank mRNA | AK037228.1 | - | 1 - 2 | 180 | 64 |