The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177723.4 | NP_808391.2 | 5 - 6 | 909 | 64 |
NR_027644.1 | 4 - 5 | 628 | 64 | ||
GenBank mRNA | AK037228.1 | - | 5 - 6 | 910 | 64 |
BC064106.1 | - | 4 - 5 | 628 | 64 |