The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177750.6 | NP_808418.1 | 5 - 6 | 656 | 93 |
GenBank mRNA | AK047241.1 | - | 3 - 4 | 2376 | 93 |
BC117734.1 | - | 3 - 4 | 312 | 93 | |
BC117735.1 | - | 3 - 4 | 312 | 93 | |
BC129957.1 | - | 3 - 4 | 312 | 93 | |
BC129958.1 | - | 3 - 4 | 312 | 93 |