The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177806.3 | NP_808474.2 | 12 - 13 | 1993 | 118 |
GenBank mRNA | AK154170.1 | - | 13 - 14 | 2617 | 118 |
AK168462.1 | - | 12 - 13 | 1989 | 118 | |
BC029153.1 | - | 7 - 8 | 965 | 118 | |
BC030904.1 | - | 12 - 13 | 1932 | 118 | |
BC033575.1 | - | 7 - 8 | 4060 | 118 | |
BC034078.1 | - | 15 - 16 | 2250 | 118 | |
BC076622.1 | - | 12 - 13 | 1679 | 118 |