Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a48
Entrez Gene ID
328258
Gene Name
solute carrier family 25, member 48
Gene Aliases
AU042651, E230025K15
Chromosome Location
Chr.13: 56437963 - 56472367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177809.4 NP_808477.2 3 - 4 297 90
GenBank mRNA AK054181.1 - 3 - 4 298 90
BC107250.1 - 3 - 4 261 90
BC107251.1 - 3 - 4 261 90

View Full Product Details