The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177822.3 | NP_808490.1 | 5 - 6 | 692 | 62 |
GenBank mRNA | AK028891.1 | - | 5 - 6 | 692 | 62 |
BC052484.1 | - | 4 - 5 | 428 | 62 |