The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163073.1 | NP_001156545.1 | 5 - 6 | 716 | 126 |
NM_172153.3 | NP_742165.1 | 5 - 6 | 729 | 126 | |
NM_178142.5 | NP_835278.2 | 5 - 6 | 716 | 126 | |
GenBank mRNA | AB076078.1 | - | 5 - 6 | 728 | 126 |
AK041987.1 | - | 5 - 6 | 675 | 126 | |
AK042567.1 | - | 5 - 6 | 730 | 126 | |
AK043498.1 | - | 5 - 6 | 674 | 126 | |
AK043845.1 | - | 5 - 6 | 684 | 126 | |
AK154398.1 | - | 5 - 6 | 677 | 126 | |
AK155421.1 | - | 5 - 6 | 677 | 126 | |
BC138563.1 | - | 5 - 6 | 716 | 126 |