Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcfl5
Entrez Gene ID
277353
Gene Name
transcription factor-like 5 (basic helix-loop-helix)
Gene Aliases
AV260129, Figlb
Chromosome Location
Chr.2: 180621956 - 180642691 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK132721.1 - 3 - 4 721 88
AY234363.1 - 3 - 4 855 88

View Full Product Details