Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a40
Entrez Gene ID
319653
Gene Name
solute carrier family 25, member 40
Gene Aliases
B230315F11Rik
Chromosome Location
Chr.5: 8422838 - 8454840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289595.1 NP_001276524.1 7 - 8 843 90
NM_001289596.1 NP_001276525.1 8 - 9 896 90
NM_178766.5 NP_848881.2 8 - 9 915 90
GenBank mRNA AK044260.1 - - 916 90
AK045863.1 - 8 - 9 903 90
AK079538.1 - 8 - 9 845 90
AK084797.1 - 7 - 8 783 90
AK088444.1 - 8 - 9 881 90
AK136770.1 - 8 - 9 877 90
AK163037.1 - 8 - 9 2630 90
BC083103.1 - 7 - 8 693 90

View Full Product Details