Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sbf2
Entrez Gene ID
319934
Gene Name
SET binding factor 2
Gene Aliases
4833411B01Rik, AA414977, AI317167, B430219L04Rik, Mtmr13, mMTMH1
Chromosome Location
Chr.7: 110308011 - 110614969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177324.2 NP_796298.2 12 - 13 1456 128
GenBank mRNA AK046622.1 - 12 - 13 1444 128
AK048952.1 - 12 - 13 1433 128
AK080952.1 - 12 - 13 1457 128
AK080963.1 - 12 - 13 1451 128
AK087396.1 - 12 - 13 1451 128
AK173257.1 - 12 - 13 1415 128
BC067204.1 - 13 - 14 1494 128

View Full Product Details