The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178415.1 | NP_848502.1 | 17 - 18 | 2183 | 69 |
NM_181316.4 | NP_851833.2 | 16 - 17 | 2127 | 69 | |
GenBank mRNA | AK028200.1 | - | 16 - 17 | 2217 | 69 |
AK053503.1 | - | 1 - 2 | 872 | 69 | |
AK086114.1 | - | 16 - 17 | 2099 | 69 | |
AK144730.1 | - | 17 - 18 | 2154 | 69 | |
AK166297.1 | - | 16 - 17 | 2108 | 69 | |
BC030419.1 | - | 14 - 15 | 1491 | 69 | |
BC046426.1 | - | 16 - 17 | 2139 | 69 | |
BC054441.1 | - | 5 - 6 | 386 | 69 |