Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wee1
Entrez Gene ID
22390
Gene Name
WEE 1 homolog 1 (S. pombe)
Gene Aliases
Wee1A
Chromosome Location
Chr.7: 110121700 - 110143299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009516.3 NP_033542.2 7 - 8 1675 101
GenBank mRNA AK011212.1 - 7 - 8 1451 101
AK039572.1 - 7 - 8 991 101
AK184246.1 - 2 - 3 154 101
AK215663.1 - 2 - 3 154 101
BC006852.1 - 7 - 8 1694 101

View Full Product Details